Learn More
American Research Products Inc PrEST Antigen C19orf60
Supplier:  American Research Products Inc APREST82438
Encompass
PrEST Antigen C19orf60 antigen APrEST82438 for chromosome 19 open reading frame 60 Recombinant protein fragment of Human C19orf60
Catalog No. 50-008-15963