Learn More
American Research Products Inc PrEST Antigen COLGALT1
Supplier:  American Research Products Inc APREST82677
Encompass
PrEST Antigen COLGALT1 antigen APrEST82677 for collagen beta(1-O)galactosyltransferase 1 Recombinant protein fragment of Human COLGALT1
Catalog No. 50-008-16201