Learn More
American Research Products Inc PrEST Antigen PTRF
Supplier:  American Research Products Inc APREST83755
Encompass
PrEST Antigen PTRF antigen APrEST83755 for polymerase I and transcript release factor Recombinant protein fragment of Human PTRF
Catalog No. 50-008-17125