Learn More
American Research Products Inc PrEST Antigen SLC16A4
Supplier:  American Research Products Inc APREST83694
Encompass
PrEST Antigen SLC16A4 antigen APrEST83694 for solute carrier family 16 member 4 Recombinant protein fragment of Human SLC16A4
Catalog No. 50-008-15954