Learn More
Medchemexpress LLC HY-10844 10mg Medchemexpress, Pretomanid CAS:187235-37-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1084410MG
Encompass_Preferred
Medchemexpress, HY-10844 10mg Pretomanid CAS:187235-37-6 Pretomanid (PA-824) is a small-molecule nitroimidazopyran drug candidate for the treatment of tuberculosis; the MIC values of PA-824 against a panel of MTB pan-sensitive and rifampin mono-resistant clinical isolates ranged from 0.015 to 0.25 ug/ml.IC50 value: 0.015 to 0.25 ug/ml (MICs) Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8550