Learn More
TARGETMOL CHEMICALS INC PRETOMANID 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T609650MG
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pretomanid ((S)-PA 824) a nitroimidazole is used as an anti-tuberculosis drug for tuberculosis with MIC less than 2.8 uM. purity: 99%
Catalog No. 50-257-6602