Learn More
Medchemexpress LLC Prexasertib (dihydrochloride) | 1234015-54-3 | 99.4% | 438.31 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY18174A5MG
Encompass_Preferred
Prexasertib dihydrochloride (LY2606368 dihydrochloride) is a selective, ATP-competitive second-generation checkpoint kinase 1 (CHK1) inhibitor with a Ki of 0.9 nM and an IC50 of <1 nM. It also inhibits CHK2 (IC50=8 nM) and RSK1 (IC50=9 nM). This compound causes double-stranded DNA breakage and replication catastrophe leading to apoptosis and shows potent anti-tumor activity.
  • Selective ATP-competitive second-generation checkpoint kinase 1 (CHK1) inhibitor.
  • Inhibits CHK2 and RSK1.
  • Causes double-stranded DNA breakage and replication catastrophe.
  • Leads to apoptosis and potent anti-tumor activity.
  • Induces growth inhibition in tumor xenografts.
  • Inhibits CHK1 in blood.
  • Promotes phosphorylation of H2AX (S139) and RPA2 (S4/S8).
  • Associated with clinical trials for various neoplasms.
Catalog No. 50-003-86825