Learn More
Selleck Chemical LLC Prexasertib HCl (LY2606368) 5mg 1234015-54-3
SDP
Supplier:  Selleck Chemical LLC S71785MG
Encompass_Preferred
Prexasertib (LY2606368) is an ATP-competitive CHK1 inhibitor with a Ki value of 0.9 nmol/L. For CHK2 and RSK, its IC50 values are 8 nM and 9 nM respectively in cell-free assay. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4493