Learn More
Cayman Chemical PRfluoroheptnsulfonIc ACD 10mg
SDP
Supplier:  Cayman Chemical 3724610 MG
Encompass_Preferred
A perfluoroalkyl substance; induces malformations in zebrafish larvae (EC50 = 168.1 µM); has been found in landfill leachate; prenatal exposure to PFHpS is associated with decreased birth weight
Catalog No. 50-291-5361