Learn More
Cayman Chemical PRfluoropentnsulfonIc ACD 10mg
SDP
Supplier:  Cayman Chemical 3725310 MG
Encompass_Preferred
A perfluoroalkyl substance; has been found as a contaminant in wastewater and breastmilk
Catalog No. 50-291-5396