Learn More
Neuromics, Inc Primary Human Neurons, 300,000 Cells are initiated by digestion of minced brain cortical tissue.
Supplier:  Neuromics, Inc HNC001
Encompass
Human Primary Neurons are initiated by digestion of minced brain cortical tissue with collagenase. Human neurons are separated from the mixture of cell populations and offered at passage 3 in a frozen vial. HNCs Growth Medium (containing 10% serum) and growth supplements, cat# HNM001 is recommended for cell culture.
Catalog No. NC2071963