Learn More
Medchemexpress LLC Pridopidine | 346688-38-8 | MFCD09835586 | 99.9% | 281.41 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY10684
Encompass_Preferred
Pridopidine is identified as a dopamine (DA) stabilizer that functions as a low-affinity dopamine D2 receptor (D2R) antagonist. It exhibits high affinity for the sigma 1 receptor (S1R), with a Ki value between 70 and 80 nM, which is approximately 100 times greater than its affinity for D2R.
  • Dopamine (DA) stabilizer
  • Low affinity dopamine D2 receptor (D2R) antagonist
  • High affinity towards sigma 1 receptor (S1R)
Catalog No. 50-003-47690