Learn More
Apexbio Technology LLC Prilocaine hydrochloride 1786-81-8 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B12805.1
Encompass_Preferred
Prilocaine hydrochloride (CAS 1786-81-8) is a local anesthetic belonging to the amino amide class It functions by reversibly inhibiting voltage-gated sodium channels on neuronal cell membranes thereby blocking the initiation and propagation of nerve impulses This property makes it suitable for investigating neuronal excitability pain signaling and ion channel pharmacology in various preclinical models Prilocaine hydrochloride is commonly employed in studies requiring local anesthesia and serves as a reference compound for benchmarking novel anesthetic agents in research settings
Catalog No. 50-000-88205