Learn More
TARGETMOL CHEMICALS INC PRIM-O-GLUCOSYLCIMIFUGIN 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T38055MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Prim-O-glucosylcimifugin (Cimifugin 7-glucoside) can inhibit the proliferation of SMC(smooth muscle cell) stimulated by TNF-alpha increase the proportion of G0/G1 phase. purity: 99%
Catalog No. 50-257-6871