Learn More
Cayman Chemical PrImaquInphosphate 1g
SDP
Supplier:  Cayman Chemical 182231 G
Encompass_Preferred
An antimalarial drug used in the treatment of persistent liver forms of P. vivax or P. ovale infection for its ability to kill late-stage gametocytes and hypnozoites
Catalog No. 50-296-6810