Learn More
Biotium Primary Antibody Biotin rBN-34 CF488A 1/EA
Supplier:  Biotium P033488A1ML
Encompass_Preferred
Biotin (rBN-34) is a recombinant mouse monoclonal antibody that recognizes free biotin and biocytin This CF488A antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-005-00706