Learn More
Biotium Primary Antibody c Fos r2H2 CF488A 1/EA
Supplier:  Biotium P043488A500UL
Encompass_Preferred
c-Fos (r2H2) is a recombinant mouse monoclonal antibody that recognizes c-Fos This CF488A antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-005-01319