Learn More
Biotium Primary Antibody c Fos r2H2 CF568 1/EA
Supplier:  Biotium P043568500UL
Encompass_Preferred
c-Fos (r2H2) is a recombinant mouse monoclonal antibody that recognizes c-Fos This CF568 antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-004-99687