Learn More
Biotium Primary Antibody CD63 24MS05.63 CF488A 1/EA
Supplier:  Biotium P020488A500
Encompass_Preferred
CD63 (24MS05 63) is a recombinant rat monoclonal antibody that recognizes the mouse tetraspanin protein CD63 This CF488A antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 100 tests
Catalog No. 50-004-97618