Learn More
Biotium Primary Antibody CD63 P08962 CF568 1/EA
Supplier:  Biotium P004568ST500
Encompass_Preferred
ExoBrite STORM CF568 CD63 Antibody is designed for detection of extracellular vesicles by STORM CF568 has been validated and recommended for STORM microscopy and has Ex/Em at 562/584 nm 500 uL
Catalog No. 50-004-96742