Learn More
Biotium Primary Antibody CD81 2304.81 CF568 1/EA
Supplier:  Biotium P014568500
Encompass_Preferred
CD81 (2304 81) is a recombinant mouse monoclonal antibody that recognizes the human tetraspanin protein CD81 This CF568 antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 100 tests
Catalog No. 50-004-96782