Learn More
Biotium Primary Antibody CD81 2304.81 CF700 1/EA
Supplier:  Biotium P014700125
Encompass_Preferred
CD81 (2304 81) is a recombinant mouse monoclonal antibody that recognizes the human tetraspanin protein CD81 This CF700 antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 25 tests
Catalog No. 50-004-97712