Learn More
Biotium Primary Antibody CD81 NA CF568 1/EA
Supplier:  Biotium P019568ST500
Encompass_Preferred
ExoBrite STORM CF568 CD81 (Mouse/Rat) Antibody is designed for detection of extracellular vesicles by STORM CF568 has been validated and recommended for STORM microscopy and has Ex/Em at 562/583 nm 500 uL
Catalog No. 50-004-97539