Learn More
Biotium Primary Antibody CD81 NA ExoBrite 410 450 1/EA
Supplier:  Biotium P019410125
Encompass_Preferred
ExoBrite 410/450 CD81 anti-mouse Flow Antibody is optimized for detection of the exosome marker CD81 in isolated exosomes by flow cytometry The conjugate has Ex/Em at 408/452 nm for detection in the Pacific Blue channel Recommended for instruments optimized for small particle flow 25 tests
Catalog No. 50-004-97617