Learn More
Biotium Primary Antibody CD81 NA ExoBrite APC 1/EA
Supplier:  Biotium P019APC125
Encompass_Preferred
ExoBrite APC CD81 anti-mouse Flow Antibody is optimized for detection of the exosome marker CD81 in isolated exosomes by flow cytometry Recommended for instruments optimized for small particle flow 25 tests
Catalog No. 50-004-97540