Learn More
Biotium Primary Antibody CD8a SK1 RPE Astral775 1/EA
Supplier:  Biotium P009R775500
Encompass_Preferred
CD8a (clone SK1) is a mouse monoclonal antibody that recognizes the human protein CD8 CD8a is a common marker of cytotoxic (Killer) T cells and is a T cell coreceptor This RPE-Astral775 antibody conjugate has been validated in flow cytometry
Catalog No. 50-004-97244