Learn More
Biotium Primary Antibody CD9 2310.9 CF647Plus 1/EA
Supplier:  Biotium P015647P125
Encompass_Preferred
CD9 (2310 9) is a recombinant mouse monoclonal antibody that recognizes the human tetraspanin protein CD9 This CF647Plus antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 25 tests
Catalog No. 50-004-99102