Learn More
Biotium Primary Antibody CD9 24MS04.9 CF405M 1/EA
Supplier:  Biotium P016405M500
Encompass_Preferred
CD9 (24MS04 9) is a recombinant rat monoclonal antibody that recognizes the mouse tetraspanin protein CD9 This CF405M antibody conjugate has been validated in flow cytometry It is supplied at 5 uL/test with sufficient antibody for 100 tests
Catalog No. 50-004-99047