Learn More
Biotium Primary Antibody CD9 P21926 CF498 1/EA
Supplier:  Biotium P003498ST500
Encompass_Preferred
ExoBrite STORM CF498 CD9 Antibody is designed for detection of extracellular vesicles by STORM CF498 has been validated and recommended for STORM microscopy and has Ex/Em at 498/518 nm 500 uL
Catalog No. 50-004-98517