Learn More
Biotium Primary Antibody CD9 P21926 CF647Plus 1/EA
Supplier:  Biotium P003647PST500
Encompass_Preferred
ExoBrite STORM CF647Plus CD9 Antibody is designed for detection of extracellular vesicles by STORM CF647Plus has been validated and recommended for STORM microscopy and has Ex/Em at 652/668 nm 500 uL
Catalog No. 50-004-97353