Learn More
Biotium Primary Antibody CHGA Chromogranin A PHE5 CF740 1/EA
Supplier:  Biotium BNC740461500
Encompass_Preferred
Chromogranin A is present in neuroendocrine cells throughout the body including the neuroendocrine cells of the large and small intestine adrenal medulla and pancreatic islets It is an excellent marker for carcinoid tumors pheochromocytomas paragangliomas and other neuroendocrine tumors Co-expression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms Reportedly co-expression of certain keratins and chromogranin indicates neuroendocrine lineage The presence of strong anti-chromogranin staining and absence of anti-keratin staining should raise the possibility of paraganglioma The co-expression of chromogranin and NSE is typical of neuroendocrine neoplasms Most pituitary adenomas and prolactinomas readily express chromogranin Primary antibodies are available purified or with a selection of fluorescent CF Dyes and other labels CF Dyes offer exceptional brightness and photostability Note Conjugates of blue fluorescent dyes like CF405S and CF405M are
Catalog No. 50-004-98076