Learn More
Biotium Primary Antibody DNP rLO-DNP-2 CF488A 1/EA
Supplier:  Biotium P038488A1ML
Encompass_Preferred
DNP (rLO-DNP-2) is a recombinant rat monoclonal antibody that recognizes the dinitrophenol (DNP) hapten This CF488A antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-005-01386