Learn More
Biotium Primary Antibody Histone H2A H2B SdAb2511.HISTONE CF488A 1/EA
Supplier:  Biotium N008488A1ML
Encompass_Preferred
Histone VHH (SdAb2511 HISTONE) is a recombinant Alpaca VHH single-domain antibody that recognizes Histone H2A/H2B with broad species reactivity Featuring Biotiums bright and photostable green fluorescent CF488A Dye this VHH is validated for immunofluorescence Supplied as 1 mL at 0 1 mg/mL
Catalog No. 50-004-98607