Learn More
Biotium Primary Antibody Human IgG Fc N2508.HFC CF770 1/EA
Supplier:  Biotium N007770500UL
Encompass_Preferred
Anti-Human IgG (Fc) rVHH (N2508 HFC) is a recombinant Alpaca VHH antibody that recognizes the Fc domain of human IgG and is validated for IF and flow Near-IR STORM-compatible CF770 dye has Ex/Em at 770/797 nm Unit size 500 uL at 0 1 mg/mL
Catalog No. 50-004-98582