Learn More
Biotium Primary Antibody Human IgG IG266 CF740 1/EA
Supplier:  Biotium BNC740266100
Encompass_Preferred
Recognizes a protein of 75 kDa identified as heavy chain of human immunoglobulins It does not cross-react with plus or minus (IgA) (IgM) (IgE) or (IgD) heavy chains T-cells monocytes granulocytes or erythrocytes This MAb is useful in the identification of leukemias plasmacytomas and certain non-Hodgkins lymphomas The most common feature of these malignancies is the restricted expression of a single heavy chain class Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant x000D x000D Primary antibodies are available purified or with a selection of fluorescent CF Dyes and other labels CF Dyes offer exceptional brightness and photostability Note Conjugates of blue fluorescent dyes like CF405S and CF405M are not recommended for detecting low abundance targets because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors x000D x000D
Catalog No. 50-004-98288