Learn More
Biotium Primary Antibody Mouse IgG1 Fc N2504.M1FC CF488A 1/EA
Supplier:  Biotium N004488A500UL
Encompass_Preferred
Anti-Mouse IgG1 (Fc) rVHH (N2504 M1FC) is a recombinant Alpaca VHH antibody that recognizes the Fc fragment of mouse IgG1 and is validated for IF flow and WB Green fluorescent STORM-compatible CF488A dye has Ex/Em at 490/515 nm Unit size 500 uL at 0 1 mg/mL
Catalog No. 50-004-98597