Learn More
Biotium Primary Antibody Mouse IgG1 Fc N2504.M1FC CF640R 1/EA
Supplier:  Biotium N004640R500UL
Encompass_Preferred
Anti-Mouse IgG1 (Fc) rVHH (N2504 M1FC) is a recombinant Alpaca VHH antibody that recognizes the Fc fragment of mouse IgG1 and is validated for IF flow and WB Far-red fluorescent photostable CF640R dye has Ex/Em at 642/662 nm Unit size 500 uL at 0 1 mg/mL
Catalog No. 50-004-98197