Learn More
Biotium Primary Antibody NeuN rA60 CF488A 1/EA
Supplier:  Biotium P037488A500UL
Encompass_Preferred
NeuN (rA60) is a recombinant mouse monoclonal antibody that recognizes neuron-specific protein NeuN This CF488A antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-004-99483