Learn More
Biotium Primary Antibody Neurofilament H rRmdO-20 CF568 1/EA
Supplier:  Biotium P035568500UL
Encompass_Preferred
Neurofilament-H (rRmdO-20) is a recombinant mouse monoclonal antibody that recognizes Neurofilament-H This CF568 antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-005-00802