Learn More
Biotium Primary Antibody Nuclear membrane 2406.NM CF640R 1/EA
Supplier:  Biotium P023640R1ML
Encompass_Preferred
This recombinant monoclonal mouse antibody conjugate stains the nuclear membrane in human cells Featuring Biotiums bright and photostable far-red fluorescent CF640R Dye (Ex/Em 642/662 nm) 100 ug/mL 1 mL
Catalog No. 50-004-97622