Learn More
Biotium Primary Antibody Nucleolin 2507.NCL CF488A 1/EA
Supplier:  Biotium P039488A500UL
Encompass_Preferred
Nucleolin (2507 NCL) is a recombinant mouse monoclonal antibody that recognizes nucleolin This CF488A antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-004-99740