Learn More
Biotium Primary Antibody Rabbit IgG Fc N2507.RFC CF488A 1/EA
Supplier:  Biotium N006488A500UL
Encompass_Preferred
Alpaca Anti-Rabbit IgG (Fc) rVHH (N2507 RFC) is a recombinant Alpaca VHH antibody that recognizes the Fc fragment of rabbit IgG and is validated for immunofluorescence Green fluorescent STORM-compatible CF488A dye has Ex/Em at 490/515 nm Unit size 500 uL at 0 1 mg/mL
Catalog No. 50-004-97497