Learn More
Biotium Primary Antibody SYT1 cytoplasmic domain SdAb2501.SYT1 CF568 1/EA
Supplier:  Biotium N002568200UL
Encompass_Preferred
SYT1 VHH (SdAb2501 SYT1) is a recombinant Alpaca VHH antibody that recognizes single-domain rat and mouse SYT1 Featuring Biotiums bright and photostable red fluorescent CF568 Dye this antibody conjugate has been validated in Immunofluorescence It is supplied at 200 uL at 0 1 mg/mL
Catalog No. 50-004-97488