Learn More
Biotium Primary Antibody VGLUT1 SdAb2412.VGLUT1 CF568 1/EA
Supplier:  Biotium N003568200UL
Encompass_Preferred
VGLUT1 VHH (SdAb2412 VGLUT1) is a recombinant Alpaca VHH antibody that recognizes single-domain rat and mouse VGLUT1 Featuring Biotiums bright and photostable red fluorescent CF568 Dye this antibody conjugate has been validated in Immunofluorescence It is supplied at 200 uL at 0 1 mg/mL
Catalog No. 50-004-97495