Learn More
Biotium Primary Antibody ZO 1 rZO1-1A12 CF647 1/EA
Supplier:  Biotium P034647500UL
Encompass_Preferred
ZO-1 (rZO1-1A12) is a recombinant mouse monoclonal antibody that recognizes ZO-1 This CF647 antibody conjugate has been validated in Immunofluorescence CF and reg dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability
Catalog No. 50-005-00448