Learn More
ATCC Primary Bladder Epithelial (A/T/N)Cells; Normal, Human
Supplier:  ATCC PCS420010
Encompass
Primary Bladder Epithelial Cells (A/T/N); Normal, Human (BdEC) are isolated from the apex, trigone, and neck regions of the bladder. The cells are grown in Bladder Epithelial Cell Basal Medium (ATCC PCS-420-032) supplemented with Bladder Epithelial Growth Kit (ATCC PCS-420-042). ATCC Primary Human Bladder Epithelial Cells serves as an essential tool in the profiling of normal human bladder epithelial cells and utilization of a model system in the study of bladder disorders, infections, bladder reconstruction studies; and advancement of cancer research. The cells are quality tested for optimal growth over a period of at least 15 population doubling (pdl). These cells are characterized through the expression of protein markers detected by immunocytochemistry staining (ICC). The cells are alpha-smooth muscle actin negative but cytokeratin 18 positive.
Catalog No. 50-238-2748
 Restricted Item