Learn More
ATCC Primary Gingival Fibroblasts; Normal, Human,
Supplier:  ATCC PCS201018
Encompass
Primary gingival fibroblast was isolated from the gingiva. The cell has applications in human gingival fibroblasts (hGF) and could potentially be an alternative source of mesenchymal stem cells (MSC) for regenerative medicine studies as they share similar morphology, CD markers, and differentiation lineage.
Catalog No. 50-238-4175