Learn More
STEMCELL Technologies Human Peripheral Blood CD8+ T Cells, Frozen, Size: 5 x 10^6 cells
SDP
Supplier:  STEMCELL Technologies 2000164
Encompass
Primary human CD8+ T cells were isolated from peripheral blood (PB) mononuclear cells (MNCs) using negative immunomagnetic separation techniques. PB was collected using the anticoagulant acid-citrate-dextrose solution A (ACDA).Cells were obtained using Institutional Review Board (IRB)-approved consent forms and protocols.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.Browse our Frequently Asked Questions (FAQs) on Primary Cells.
Catalog No. 50-206-9411