Learn More
Waters Corp Waters Primary Inlet Check Valve Filter Kit, Stainless Steel, 2 per Pack
Supplier:  Waters Corp 700002912
Encompass_Preferred
The Stainless Steel Primary Inlet Check Valve Filter Kit is a standard component designed for use in the ACQUITY Binary Solvent Manager. This kit provides essential replacement parts to maintain optimal system performance.
  • Standard kit for ACQUITY Binary Solvent Manager.
  • Includes 2 units per package.
  • Suitable for UPC2 and UPLC system types.
  • Features a check valve type.
Catalog No. 50-203-9458