Learn More
ATCC Primary Lobar Bronchial Epithelial Cells; Normal; Human
Supplier:  ATCC PCS300015
Encompass
Normal Human Primary Lobar Epithelial Cells were isolated from the respiratory tract of a donor and when grown in Airway Epithelial Cell Basal Media supplemented with Bronchial/Tracheal Epithelial Cell Growth Kit components, provide an ideal cell system to propagate bronchial/tracheal epithelial cells in serum-free conditions.
Catalog No. 50-238-4701